![4-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/155/261/7c90c3b0-b083-4f09-bdb7-01a5fea9e5ae/640/7c90c3b0-b083-4f09-bdb7-01a5fea9e5ae.png)
4-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
![4-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/155/261/7c90c3b0-b083-4f09-bdb7-01a5fea9e5ae/640/7c90c3b0-b083-4f09-bdb7-01a5fea9e5ae.png)
Empirical Formula (Hill Notation):
C5H8Cl2N2
CAS Number:
Molecular Weight:
167.04
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing