跳轉至內容
Merck
  • Hepatic expression of malonyl-CoA decarboxylase reverses muscle, liver and whole-animal insulin resistance.
Hepatic expression of malonyl-CoA decarboxylase reverses muscle, liver and whole-animal insulin resistance.
Nature medicine (2004-02-11)
Jie An, Deborah M Muoio, Masakazu Shiota, Yuka Fujimoto, Gary W Cline, Gerald I Shulman, Timothy R Koves, Robert Stevens, David Millington, Christopher B Newgard
摘要
Lipid infusion or ingestion of a high-fat diet results in insulin resistance, but the mechanism underlying this phenomenon remains unclear. Here we show that, in rats fed a high-fat diet, whole-animal, muscle and liver insulin resistance is ameliorated following hepatic overexpression of malonyl-coenzyme A (CoA) decarboxylase (MCD), an enzyme that affects lipid partitioning. MCD overexpression decreased circulating free fatty acid (FFA) and liver triglyceride content. In skeletal muscle, levels of triglyceride and long-chain acyl-CoA (LC-CoA)-two candidate mediators of insulin resistance-were either increased or unchanged. Metabolic profiling of 36 acylcarnitine species by tandem mass spectrometry revealed a unique decrease in the concentration of one lipid-derived metabolite, beta-OH-butyrate, in muscle of MCD-overexpressing animals. The best explanation for our findings is that hepatic expression of MCD lowered circulating FFA levels, which led to lowering of muscle beta-OH-butyrate levels and improvement of insulin sensitivity.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
硬脂酰-L-肉碱, ≥97.0% (TLC)