跳轉至內容
Merck
  • Potential chemoprevention of diethylnitrosamine-induced hepatocarcinogenesis in rats: myrrh (Commiphora molmol) vs. turmeric (Curcuma longa).
Potential chemoprevention of diethylnitrosamine-induced hepatocarcinogenesis in rats: myrrh (Commiphora molmol) vs. turmeric (Curcuma longa).
Acta histochemica (2011-08-30)
Mohamed El-Shahat, Sabah El-Abd, Mohamed Alkafafy, Gamal El-Khatib
摘要
The aim of the present study was to assess the potential chemopreventive effects of myrrh (Commiphora molmol) vs. turmeric (Curcuma longa) in hepatocarcinogenic rats induced by a single intraperitoneal injection of diethylnitrosamine (DENA) (200 mg/kg body weight). Ninety male Wistar rats used in this study were randomly divided into six equal groups (n=15). Group 1 rats served as negative controls; group 2 received a single i.p. injection of DENA and served as positive controls. Rats in both groups were fed on basal diet. Group 3 rats were fed a diet containing 5% turmeric, whereas group 4 rats were fed a diet containing 2% myrrh. Rats in groups 5 and 6 received a single i.p. injection of DENA and were fed diets containing 5% turmeric and 2% myrrh, respectively. The study demonstrated that DENA caused a significant increase in serum indices of liver enzymes and also severe histological and immunohistochemical changes in hepatic tissues. These included disorganized hepatic parenchyma, appearance of pseudoacinar and trabecular arrays of hepatocytes and alterations in CD10-immunoreactivity. Dietary supplementation of turmeric relatively improved the biochemical parameters to values approximating those of the negative controls and delayed the initiation of carcinogenesis. In contrast, myrrh did not improve the biochemical parameters or delay the hepatocarcinogenesis. Both turmeric and myrrh induced significant biochemical and histological changes in non-treated rats. In conclusion, DENA significantly changes the biological enzymatic activities in serum and the integrity of hepatic tissues. Phytochemicals with potential hepatoprotective effects must be applied cautiously owing to their potential hepatotoxicity.