跳轉至內容
Merck
  • Anti-coccidial effect of Commiphora molmol in the domestic rabbit (Oryctolagus cuniculus domesticus L.).
Anti-coccidial effect of Commiphora molmol in the domestic rabbit (Oryctolagus cuniculus domesticus L.).
Journal of the Egyptian Society of Parasitology (2011-01-28)
Hanadi B Baghdadi, Ebtesam M Al-Mathal
摘要
This study investigated the effect of oleo-gum-resin of Commiphora monlmol Engler (Family: Burceraceae) known as Myrrh and the commercial extract known as Mirazid as a treatment against hepatic coccidiosis induced by the parasite Eimeria stiedae in domestic rabbits. Rabbits were infected with 1000 parasite sporulated oocysts and subjected to two treatment regimens, using crude myrrh suspension and the oleo-resin extract, mirazid, each administered at 500 mg/kg rabbit body weight. Treatments of infected rabbits resulted in significant reduction of the mean oocyst numbers in rabbit faeces by 52.38% in the crude-treated rabbits and by 90.90% mirazid-treated rabbits, compared to the untreated infected rabbits at day 21 post-infection (pi). At day 28 pi no oocysts parasite were observed in the faeces of rabbits. Both treatments resulted in significant recovery of infected rabbits from all symptoms of infection compared to the untreated infected group and healthy control groups. Histopathological examination of liver showed remarkable improvement in all histopathological parameters in G5 and G8 compared with the infected untreated G2. These included an almost complete healing of the hemorrhagic tissue and partial healing of the endothelial lining and hepatocytes encircling the central vein, the hepatocytes laminate regained their original radial shape and disappearance of fat vacuoles from the tissue and remarkable reduction in lymphocytes infiltration, decreased hyperplasia of the epithelial cells with significant decreasing of the parasite stage numbers. Results also indicate that mirazid was more effective than crude myrrh, probably due to higher content of purified active ingredients.