跳轉至內容
Merck
  • Chiral porphyrin dimer with a macrocyclic cavity for intercalation of aromatic guests.
Chiral porphyrin dimer with a macrocyclic cavity for intercalation of aromatic guests.
Chemical communications (Cambridge, England) (2011-04-13)
Tadashi Ema, Norichika Ura, Katsuya Eguchi, Yuhei Ise, Takashi Sakai
摘要
Chiral diporphyrin receptor 1, which has a macrocyclic cavity to sandwich aromatic guest molecules via double π-π stacking interactions, enabled the naked-eye detection of an aromatic explosive as well as chiral discrimination in NMR.