跳轉至內容
Merck
  • A novel exonuclease III aided amplification method for sensitive nucleic acid detection based on single walled carbon nanotube induced quenching.
A novel exonuclease III aided amplification method for sensitive nucleic acid detection based on single walled carbon nanotube induced quenching.
Chemical communications (Cambridge, England) (2011-11-23)
Hui Chen, Jiaqin Wang, Guohai Liang, Peng Zhang, Jilie Kong
摘要
We describe herein a novel exonuclease III aided amplification method based on single walled carbon nanotube quenching (EASQ) for sensitive and convenient nucleic acid detection, which enabled 80-fold decrease of detection limit for HIV1 DNA assay compared with no target recycling.