跳轉至內容
Merck
  • Photodegradation of etridiazole by UV radiation during drinking water treatment.
Photodegradation of etridiazole by UV radiation during drinking water treatment.
Chemosphere (2009-05-30)
Chao Liu, Zhimin Qiang, Fang Tian, Tao Zhang
摘要
The photodegradation of etridiazole (ETZ) in water by UV radiation at 254 nm was investigated. Results indicate that the simulated first-order rate constants decreased with the increase of initial ETZ concentration (i.e., 5, 20 and 30 microM), and did not show any pH dependence within the range from 6.0 to 8.0. The quantum yield was 0.46+/-0.02 molE(-1) at pH 7.0. H(2)O(2) was generated at trace levels in the range from 0 to 1.0 microM during photodegradation of ETZ. Direct photodegradation was responsible for the decomposition of ETZ in distilled water by UV radiation. Three organic byproducts were identified: 5-ethoxy-3-dichloromethyl-1,2,4-thiadiazole, 5-ethoxy-1,2,4-thiadiazole-3-carboxylic acid and 5-ethoxy-3-hydroxyl-1,2,4-thiadiazole. About 90% of chloro mass in the initial ETZ was released as Cl(-) at the end of photodegradation. In contrast, the formation of sulfate and nitrate was insignificant. In general, ETZ decayed more quickly in groundwater than in sand-filtered or surface water. It is reasonably deduced that ETZ may not get removed effectively under a typical UV dose of 40 mJcm(-2) at most water treatment plants that employ UV radiation for disinfection.
材料
產品編號
品牌
產品描述
Supelco
土菌灵, PESTANAL®, analytical standard