跳轉至內容
Merck
  • Long-acting anticholinesterases for myasthenia gravis: synthesis and activities of quaternary phenylcarbamates of neostigmine, pyridostigmine and physostigmine.
Long-acting anticholinesterases for myasthenia gravis: synthesis and activities of quaternary phenylcarbamates of neostigmine, pyridostigmine and physostigmine.
Bioorganic & medicinal chemistry (2010-07-16)
Qian-Sheng Yu, Harold W Holloway, Weiming Luo, Debomoy K Lahiri, Arnold Brossi, Nigel H Greig
摘要
The N-monophenylcarbamate analogues of neostigmine methyl sulfate (6) and pyridostigmine bromide (8) together with their precursors (5), (7), and the N(1)-methylammonium analogues of (-)-phenserine (12), (-)-tolserine (14), (-)-cymserine (16) and (-)-phenethylcymserine (18) were synthesized to produce long-acting peripheral inhibitors of acetylcholinesterase or butyrylcholinesterase. Evaluation of their cholinesterase inhibition against human enzyme ex vivo demonstrated that, whereas compounds 5-8 possessed only marginal activity, 12, 14, 16 and 18 proved to be potent anticholinesterases. An extended duration of cholinesterase inhibition was determined in rodent, making them of potential interest as long-acting agents for myasthenia gravis.
材料
產品編號
品牌
產品描述
Sigma-Aldrich
新斯的明