Z804479
1 of 10
Supelco
1.18381
Calibration standard
50139-U
LiChrospher® RP-18 HPLC 色谱柱
1.08160
UV-VIS Standard 1: Potassium dichromate solution
NIST2034
Holmium Oxide Solution Wavelength Standard (240 nm to 650 nm)
58298
SUPELCOSIL™ LC-18 HPLC 柱
82011-U
Hamilton PRP-X100 HPLC色谱柱
82023-U
82039-U
Hamilton PRP-X100 HPLC Guard Cartridge
1.18349
Calibration Standard Set
54775
Hellma 667-UV35
UV/Vis spectroscopy: suitable
尋找類似的產品? 前往 產品比較指南
從最近期的版本中選擇一個:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.