文件
Z308749
Quickfit® conical flasks
capacity 100 mL, joint: ST/NS 24/29
同義詞:
FE100/3
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121804
NACRES:
NB.43
材料
borosilicate glass
conical flask
flat bottom
接合處
joint: ST/NS24/29
無菌
non-sterile
特點
closure type Standard Taper Joint
without cap
包裝
case of 5
製造商/商標名
Quickfit FE100/3
存貨情形
not available in USA and Canada
容量
100 mL
尋找類似的產品? 前往 產品比較指南
一般說明
With ground-glass socket for general use. Not suitable for vacuum use.
法律資訊
Quickfit is a registered trademark of SciLabware Limited