Merck
  • Chlamydomonas reinhardtii responding to high light: a role for 2-propenal (acrolein).
Chlamydomonas reinhardtii responding to high light: a role for 2-propenal (acrolein).
Physiologia plantarum (2017-03-23)
Thomas Roach, Theresa Baur, Wolfgang Stöggl, Anja Krieger-Liszkay
ABSTRACT
High light causes photosystem II to generate singlet oxygen (
Product Description
Supelco
Zeaxanthin, analytical standard