Merck
  • Rescue of Hearing by Gene Delivery to Inner-Ear Hair Cells Using Exosome-Associated AAV.
Rescue of Hearing by Gene Delivery to Inner-Ear Hair Cells Using Exosome-Associated AAV.
Molecular therapy : the journal of the American Society of Gene Therapy (2017-01-14)
Bence György, Cyrille Sage, Artur A Indzhykulian, Deborah I Scheffer, Alain R Brisson, Sisareuth Tan, Xudong Wu, Adrienn Volak, Dakai Mu, Panos I Tamvakologos, Yaqiao Li, Zachary Fitzpatrick, Maria Ericsson, Xandra O Breakefield, David P Corey, Casey A Maguire
ABSTRACT
Adeno-associated virus (AAV) is a safe and effective vector for gene therapy for retinal disorders. Gene therapy for hearing disorders is not as advanced, in part because gene delivery to sensory hair cells of the inner ear is inefficient. Although AAV transduces the inner hair cells of the mouse cochlea, outer hair cells remain refractory to transduction. Here, we demonstrate that a vector, exosome-associated AAV (exo-AAV), is a potent carrier of transgenes to all inner ear hair cells. Exo-AAV1-GFP is more efficient than conventional AAV1-GFP, both in mouse cochlear explants in vitro and with direct cochlear injection in vivo. Exo-AAV shows no toxicity in vivo, as assayed by tests of auditory and vestibular function. Finally, exo-AAV1 gene therapy partially rescues hearing in a mouse model of hereditary deafness (lipoma HMGIC fusion partner-like 5/tetraspan membrane protein of hair cell stereocilia [Lhfpl5/Tmhs
Sigma-Aldrich
Calix[8]arene, technical, ≥90% (CH)