Merck
  • Investigation of variants in the promoter region of PIK3C3 in schizophrenia.
Investigation of variants in the promoter region of PIK3C3 in schizophrenia.
Neuroscience letters (2008-04-19)
Ruqi Tang, Xinzhi Zhao, Chao Fang, Wei Tang, Ke Huang, Lei Wang, Huafang Li, Guoyin Feng, Shaomin Zhu, Huijun Liu, Lin He, Yongyong Shi
ABSTRACT
The PIK3C3 gene has been implicated as a candidate gene for schizophrenia by functional evidence and genetic association studies. A series of previous studies have found susceptibility SNPs in promoter region. To further verify its susceptibility to schizophrenia in the Chinese population and the function of the polymorphisms, we performed a case control study in 556 unrelated schizophrenia patients and 563 normal controls as well as an in vitro functional analysis. In our association analysis of-432C-/T, we discovered obvious differences in allele frequency between patients and controls (P=0.017). A T/C haplotype constructed by -432C-/T and -86insC, which are tightly linked with each other (r(2)=1) can significantly weaken promoter's transcriptional activity by 20% (p=0.002 by t-test). Though we cannot exclude the possibility that susceptibility of -432C-/T is caused by its linkage disequilibrium with other causal variants, our results do support PIK3C3 play a significant role in the etiology of schizophrenia.