Merck
  • A novel Crumbs3 isoform regulates cell division and ciliogenesis via importin beta interactions.
A novel Crumbs3 isoform regulates cell division and ciliogenesis via importin beta interactions.
The Journal of cell biology (2007-07-25)
Shuling Fan, Vanessa Fogg, Qian Wang, Xiao-Wei Chen, Chia-Jen Liu, Ben Margolis
ABSTRACT
The Crumbs family of apical transmembrane proteins regulates apicobasal polarity via protein interactions with a conserved C-terminal sequence, ERLI. However, one of the mammalian Crumbs proteins, Crumbs3 (CRB3) has an alternate splice form with a novel C-terminal sequence ending in CLPI (CRB3-CLPI). We report that CRB3-CLPI localizes to the cilia membrane and a membrane compartment at the mitotic spindle poles. Knockdown of CRB3-CLPI leads to both a loss of cilia and a multinuclear phenotype associated with centrosomal and spindle abnormalities. Using protein purification, we find that CRB3-CLPI interacts with importin beta-1 in a Ran-regulated fashion. Importin beta-1 colocalizes with CRB3-CLPI during mitosis, and a dominant-negative form of importin beta-1 closely phenocopies CRB3-CLPI knockdown. Knockdown of importin beta-1 blocks targeting of CRB3-CLPI to the spindle poles. Our data suggest an expanded role for Crumbs proteins in polarized membrane targeting and cell division via unique interactions with importin proteins.