Merck
An efficient synthetic approach to 6,5'-(S)- and 6,5'-(R)-cyclouridine.
Chemical communications (Cambridge, England) (2012-04-26)
Christopher S Theile, Larry W McLaughlin
ABSTRACT
Here we present new routes for the efficient syntheses of 6,5'-(S)- and 6,5'-(R)-cyclouridine. The syntheses utilize readily accessible uridine as a starting material. This route to the R diastereomer is significantly more efficient than previous synthetic efforts, allowing us to obtain large amounts of pure material for future biological testing.