Merck
[Experimental modelling of toxic encephalopathy].
Meditsina truda i promyshlennaia ekologiia (2008-10-24)
L M Sosedova, N L Iakimova, E A Titov, E A Kapustina
ABSTRACT
The article covers problem of experimental biologic modelling of toxic encephalopathies. The authors represent major methodic approaches to modelling, criteria and methods to evaluate pathologic condition of CNS, present results of own research in creating models of toxic encephalopathies caused by metallic mercury and vinylchloride inhalation.
Sigma-Aldrich
Lead(II) sulfate, 98%
Sigma-Aldrich
Lead(II) sulfate, 99.995% trace metals basis