Merck
Regio- and enantioselectivity of soybean fatty acid epoxide hydrolase.
The Journal of biological chemistry (1992-06-15)
E Blée, F Schuber
ABSTRACT
Soluble epoxide hydrolase purified from soybean catalyzes trans-addition of water across the oxirane ring of cis-9,10-epoxystearic acid with inversion of configuration at the attacked carbon, yielding threo-9,10-dihydroxystearic acid. Kinetic analyses of the progress curves, obtained at low substrate concentrations (i.e. [S] much less than Km), and determination of the enantiomeric excess of the residual substrate by chiral-phase high-performance liquid chromatography at different reaction times, indicate that the epoxide hydrolase hydrates preferentially cis-9R, 10S-epoxystearic acid (V/Km ratio, approximately 20). Interestingly, this enantiomer is obtained by epoxidation of oleic acid catalyzed by peroxygenase, a hydroperoxide-dependent oxidase, we have previously described in soybean (Blée, E., and Schuber, F. (1990) J.Biol. Chem. 265, 12887-12894). For the epoxide hydrolase to show high enantioselectivity there must be a free carboxylic acid functionality on the substrate which probably influences its positioning within the active site. This selectivity, which in principle can be used for kinetic resolution of the cis-9,10-epoxystearic acid enantiomers, is much reduced with methyl cis-9,10-epoxystearate. 18O-Labeling experiments indicate that water attacks both cis-9,10-epoxystearic acid enantiomers on the oxirane carbon which has the S-chirality. Results show that soybean epoxide hydrolase produces exclusively threo-9R,10R-dihydroxystearic acid, i.e. a naturally occurring metabolite in higher plants. cis-9,10-Epoxy-18-hydroxystearic acid, a cutin monomer, was a poorer substrate of the epoxide hydrolase than 9,10-epoxystearic acid (V/Km ratio for the preferred enantiomers, approximately 19). From a physiological point of view, peroxygenase and this newly described epoxide hydrolase could be responsible, in vivo, for the biosynthesis of a class of oxygenated fatty acid compounds known to be involved in cutin monomers production and in plant defense mechanisms.