Merck
  • Quantifying fenobucarb residue levels in beef muscles using liquid chromatography-tandem mass spectrometry and QuEChERS sample preparation.
Quantifying fenobucarb residue levels in beef muscles using liquid chromatography-tandem mass spectrometry and QuEChERS sample preparation.
Food chemistry (2013-03-19)
Ki Hun Park, Jeong-Heui Choi, A M Abd El-Aty, Md Musfiqur Rahman, Jin Jang, Ah-Young Ko, Ki Sung Kwon, Hee Ra Park, Hyung Soo Kim, Jae-Han Shim
ABSTRACT
This paper describes a comparison of the properties of the three versions of the QuEChERS method (quick, easy, cheap, effective, rugged and safe) - the original (unbuffered), acetate-buffered, and citrate-buffered methods - for the determination of fenobucarb residues in beef muscles via liquid chromatography-electrospray ionisation tandem mass spectrometry (LC-ESI(+)-MS/MS). The recovery results were good for all the versions; however, the acetate-buffered version gave higher and more consistent recoveries for fenobucarb than the other versions. Performance characteristics, such as linearity, accuracy, and precision were determined. Matrix-matched standard calibration was used for quantification, obtaining recoveries in the range of 83.7-93.4% with relative standard deviations of <5%, at two spiking levels: 10 and 40 μg/kg. The limits of detection (LOD) and quantification (LOQ) were estimated to be 1.5 and 5 μg/kg, respectively. Finally, the method was applied to the analysis of 15 market samples, and no residues were found over the limit of quantification. The method developed was found able to determine the analyte with satisfactory intensity and accuracy.
Product Description
Supelco
Fenobucarb, PESTANAL®, analytical standard
Pricing and availability is not currently available.