Merck
  • Epiretinal membrane after pars plana vitrectomy for primary pseudophakic or aphakic rhegmatogenous retinal detachment: incidence and outcomes.
Epiretinal membrane after pars plana vitrectomy for primary pseudophakic or aphakic rhegmatogenous retinal detachment: incidence and outcomes.
Retina (Philadelphia, Pa.) (2012-03-15)
Vicente Martínez-Castillo, Anna Boixadera, Laura Distéfano, Miguel Zapata, José García-Arumí
ABSTRACT
To compare outcomes of eyes that did or did not develop an epiretinal membrane (ERM) after repair of primary pseudophakic or aphakic rhegmatogenous retinal detachment by pars plana vitrectomy alone. Interventional, prospective, consecutive case series. Of 312 consecutive eyes that underwent pars plana vitrectomy for primary rhegmatogenous retinal detachment, 28 (8.97%) developed ERM during the postoperative period. Best-corrected visual acuity (BCVA) and fundus examinations were performed 1 month and 3, 6, and 12 months postoperatively. Eyes with an ERM more often had a break at the equator (odds ratio, 3.92; 95% confidence interval, 1.3–11.2; P < 0.011). At ERM diagnosis, BCVA ranged from 20/400 to 20/25 with a mean of 20/100 (0.75 logarithm of the minimum angle of resolution), and BCVA at the final visit ranged from 20/200 to 20/25 with a mean of 20/63 (0.53 logarithm of the minimum angle of resolution). Of the 22 eyes that underwent surgical removal of the ERM, BCVA was ≥ 20/40 in 5 (22.7%) before the surgical procedure and in 13 (59%) at the final follow-up visit. The incidence of ERM after pars plana vitrectomy alone for the repair of primary pseudophakic or aphakic rhegmatogenous retinal detachment was similar to reported incidences after scleral buckling. Best-corrected visual acuity improved after surgical removal of ERM, although eyes with the macula detached had lower final BCVA.
Sigma-Aldrich
Perfluorooctane, 98%