- Oxamyl dipeptide caspase inhibitors developed for the treatment of stroke.
Oxamyl dipeptide caspase inhibitors developed for the treatment of stroke.
Bioorganic & medicinal chemistry letters (2004-04-28)
Steven D Linton, Teresa Aja, Peter R Allegrini, Thomas L Deckwerth, Jose-Luis Diaz, Bastian Hengerer, Julia Herrmann, Kathy G Jahangiri, Joerg Kallen, Donald S Karanewsky, Steven P Meduna, Kip Nalley, Edward D Robinson, Silvio Roggo, Giorgio Rovelli, Andre Sauter, Robert O Sayers, Albert Schmitz, Robert Smidt, Robert J Ternansky, Kevin J Tomaselli, Brett R Ullman, Christoph Wiessner, Joe C Wu
PMID15109679
ABSTRACT
Structural modifications were made to a previously described acyl dipeptide caspase inhibitor, leading to the oxamyl dipeptide series. Subsequent SAR studies directed toward the warhead, P2, and P4 regions of this novel peptidomimetic are described herein.