Merck
  • The receptor for activated C-kinase-I (RACK-I) anchors activated PKC-beta on melanosomes.
The receptor for activated C-kinase-I (RACK-I) anchors activated PKC-beta on melanosomes.
Journal of cell science (2004-07-15)
Hee-Young Park, Heng Wu, Christina E Killoran, Barbara A Gilchrest
ABSTRACT
Protein kinase C (PKC), a family of at least eleven isoforms, mediates numerous cell functions. In human melanocytes, alpha, beta, delta, epsilon and zeta isoforms of PKC are expressed, but uniquely PKC-beta activates tyrosinase, the key and the rate-limiting enzyme in melanogenesis, by phosphorylating specific serine residues on its cytoplasmic domain. To investigate the mechanism by which only PKC-beta phosphorylates tyrosinase, we examined the expression of receptor for activated C-kinase-I (RACK-I), a receptor specific for activated PKC-beta, on the surface of melanosomes, the specialized organelle in which melanogenesis occurs. Immunoblot analysis of purified melanosomes revealed that RACK-I is readily detectable. Immunoprecipitation of RACK-I from purified melanosomes, followed by immunoblot analysis using antibody against PKC-beta, revealed abundant PKC-beta, whereas PKC-alpha was not detected when immunoblot analysis was performed using antibody against PKC-alpha. Activation of PKC in melanocytes increased the level of PKC-beta co-immunoprecipitated with RACK-I, while the level of melanosome-associated RACK-I decreased when melanocytes were treated chronically with the 12-0-tetradecanoyl-phorbol 13-Acetate (TPA), a condition known to deplete PKC and reduce tyrosinase activity. Immunoprecipitation with RACK-I antibody co-precipitated fewer PKC-beta in the presence of UV-activated 1, 1'-decamethylenebis-4-aminoquinaldinium di-iodide (DECA), known to disrupt the interaction between activated PKC-beta and RACK-I. Treatment of intact melanocytes with DECA also decreased tyrosinase activity. Moreover, suppression of RACK-I expression by transfecting melanocytes with siRNA against RACK-I reduced the basal tyrosinase activity and blocked TPA-induced increases in tyrosinase activity. Taken together, these results demonstrate that RACK-I anchors activated PKC-beta on the melanosome membrane, allowing PKC-beta to phosphorylate tyrosinase.