Merck
Microbiology of the gut of the kola nut weevil, Balanogastris kolae.
Journal of insect science (Online) (2013-02-21)
T O Femi-Ola, A G Babalola
ABSTRACT
Reports have shown that many insects have microbes in their gut system. Gut microbes are very important for insect vitality and much of their nutrition is derived from products of microbial metabolism. The habitat of Balanogastris kolae (Desbrocher des Loges) (Coleoptera: Curculionidae) suggests that they possess the ability to digest varieties of sugars particularly starch and protein materials present in the kola nut, Cola nitida Schott & Endlicher (Malvales: Malvaceae). The aim of this study was to characterize the gut bacterial communities of the kola weevil, B. kolae. To ascertain this, the gut bacterial community of a kola nut-feeding weevil, B. kolae was characterized using culture-dependent methods. The bacterial counts in the foregut, midgut and hindgut were 7.14 ± 0.11 × 10(6)cfu ml(-1), 2.68 ± 0.13 × 10(7) cfu ml(-1) and 1.43 ± 0.20 × 10(6) cfu ml(-1) respectively. There were no significant differences in the total bacterial count of the foregut, midgut and hindgut. The bacterial species were identified to be Fusobacterium nucleatum, Staphylococcus aureus, Bacillus subtilis, Corynebacterium fascians, Arthrobacter globiformis, Serratia marcescens, Bacillus brevis, Vibrio haemolyticus and Flavobacterium breve. The majority of these isolates were demonstrated to have both proteolytic and amylolytic activities.
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid