Merck
  • Hereditary multiple osteochondromas in Jordanian patients: Mutational and immunohistochemical analysis of EXT1 and EXT2 genes.
Hereditary multiple osteochondromas in Jordanian patients: Mutational and immunohistochemical analysis of EXT1 and EXT2 genes.
Oncology letters (2021-02-09)
Ziyad Mohaidat, Khaldon Bodoor, Rowida Almomani, Mohammed Alorjani, Mohammad-Akram Awwad, Audai Bany-Khalaf, Khalid Al-Batayneh
ABSTRACT
The aim of the present study was to investigate the molecular characteristics of hereditary multiple osteochondromas (HMO) in a subset of Jordanian patients with a focus on the genetic variants of exostosin (EXT1)/(EXT2) and their protein expression. Patients with HMO and their family members were included. Recorded clinical characteristics included age, sex, tumors number and location, joint deformities and associated functional limitations. Mutational analysis of EXT1 and EXT2 exonic regions was performed. Immunohistochemical staining for EXT1 and EXT2 was performed manually using two different commercially available rabbit anti-human EXT1 and EXT2 antibodies. A total of 16 patients with HMO from nine unrelated families were included, with a mean age of 13.9 years. A total of 75% (12/16) of the patients were male and (69%) (11/16) had a mild disease (class I). EXT mutation analysis revealed only EXT1 gene mutations in 13 patients. Seven variants were detected, among which three were novel: c.1019G>A, p. (Arg340His), c.962+1G>A and c.1469del, p. (Leu490Argfs*9). Of the 16 patients, 3 did not harbor any mutations for either EXT1 or EXT2. Immunohistochemical examination revealed decreased expression of EXT1 protein in all patients with EXT1 mutation. Surprisingly, EXT2 protein was not detected in these patients, although none had EXT2 mutations. The majority of Jordanian patients with HMO, who may represent an ethnic group that is infrequently investigated, were males and had a mild clinical disease course; whereas most patients with EXT1 gene mutations were not necessarily associated with a severe clinical disease course. The role of EXT2 gene remains a subject of debate, since patients with EXT1 mutations alone did not express the non-mutated EXT2 gene.