Merck
  • Transmembrane proteins are not required for early stages of nuclear envelope assembly.
Transmembrane proteins are not required for early stages of nuclear envelope assembly.
The Biochemical journal (2006-09-07)
Corinne Ramos, Elvira R Rafikova, Kamran Melikov, Leonid V Chernomordik
ABSTRACT
All identified membrane fusion proteins are transmembrane proteins. In the present study, we explored the post-mitotic reassembly of the NE (nuclear envelope). The proteins that drive membrane rearrangements in NE assembly remain unknown. To determine whether transmembrane proteins are prerequisite components of this fusion machinery, we have focused on nuclear reconstitution in a cell-free system. Mixing of soluble interphase cytosolic extract and MV (membrane vesicles) from amphibian eggs with chromatin results in the formation of functional nuclei. We replaced MV and cytosol with protein-free phosphatidylcholine LS (liposomes) that were pre-incubated with interphase cytosol. While later stages of NE assembly yielding functional nucleus did not proceed without integral proteins of MV, LS-associated cytosolic proteins were sufficient to reconstitute membrane targeting to the chromatin and GTP-dependent lipid mixing. Binding involved LS-associated A-type lamin, and fusion involved Ran GTPase. Thus in contrast with post-fusion stages, fusion initiation in NE assembly, like membrane remodelling in budding and fission, does not require transmembrane proteins.
Sigma-Aldrich
Anti-Histone Antibody, clone H11-4, clone H11-4, Chemicon®, from mouse