- Stabilization of organometallic halide perovskite nanocrystals in aqueous solutions and their applications in copper ion detection.
Stabilization of organometallic halide perovskite nanocrystals in aqueous solutions and their applications in copper ion detection.
Chemical communications (Cambridge, England) (2018-05-22)
Chunqing Ma, Ming-Fai Lo, Chun-Sing Lee
PMID29781482
ABSTRACT
Stabilization of two-dimensional (2D) PEA2PbI4 (PEA is phenethylammonium) perovskite nanocrystals (PNCs) in water is achieved. By inhibiting the desorption process, the PNCs show exceptional stability for more than 2 months in PEA+ aqueous solutions. Stabilized PNCs are successfully applied for probing Cu2+ in aqueous solution.