Merck
  • Elevated expression of NFE2L3 predicts the poor prognosis of pancreatic cancer patients.
Elevated expression of NFE2L3 predicts the poor prognosis of pancreatic cancer patients.
Cell cycle (Georgetown, Tex.) (2018-09-11)
Hui Wang, Ming Zhan, Ruimeng Yang, Yongheng Shi, Qiang Liu, Jian Wang
ABSTRACT
The highly malignant feature and difficulties for early diagnosis are the key reasons contributing to the poor prognosis of pancreatic cancer (PC) patients. NFE2L3 is a nuclear transcription factor, which has been reported an important biomarker of several tumors. But the role of NFE2L3 in PC remained undefined. Herein, through qPCR and immunohistochemistry, we found a significantly increased NFE2L3 in PC tissues as compared with adjacent non-tumor tissues. While reducing NFE2L3 expression suppressed the invasion abilities of PC cells, elevated NFE2L3 was found associated with lymph node metastasis (P = 0.001; HR = 3.95; 95% CI: 1.70 - 9.17) and advanced TNM stages (P < 0.001; HR = 4.06; 95% CI: 1.74 - 9.46). Consistently, data from both our two cohorts and the TCGA database revealed that higher NFE2L3 PC carriers had worse outcomes than those lower NFE2L3 expressers. Lastly, we confirmed the regulatory role of NFE2L3 on VEGFA, an important player involved in tumor angiogenesis. Collectively, our investigations suggested the oncogenic role of NFE2L3 in PC development and provided the rational for future adding NFE2L3 for the risk assessment of PC patients. NFE2L3: NF-E2-related factor 3; UHMK1: U2AF homology motif kinase 1; VEGFA: vascular endothelial growth factor A; GEO: gene expression omnibus; TCGA: The Cancer Genome Atlas; HPDE: human pancreas duct cells; OS: overall survival; IHC: immunohistochemistry; FFPE: formalin-fixed and paraffin-embedded; SEM: standard error of mean.
Sigma-Aldrich
Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder