Skip to Content
Merck
S3I-201 ≥97% (HPLC)

S3I-201

S3I-201 ≥97% (HPLC)
Synonym(s):
STAT3 Inhibitor VI, S3I-201, 2-Hydroxy-4-[[[[(4-methylphenyl)sulfonyl]oxy]acetyl]amino]-benzoic acid, NSC 74859
Empirical Formula (Hill Notation):
C16H15NO7S
CAS Number:
Molecular Weight:
365.36

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing