(R)-(+)-Atenolol
Synonym(s):
(+)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
Linear Formula:
(CH3)2CHNHCH2CH(OH)CH2OC6H4CH2CONH2
CAS Number:
Molecular Weight:
266.34
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing