Skip to Content
Merck
All Photos(4)

Key Documents

CMC0011

Sigma-Aldrich

XLDNA V2 Electrocompetent cells

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

Pricing and availability is not currently available.

Product Name

XLDNA V2 Electrocompetent cells, for BAC & cosmid cloning

biological source

Escherichia coli

grade

for molecular biology

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: electrocompetent
transformation efficiency: ≥1 x 1010 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The XLDNA V2 Electrocompetent cells are designed for transformation of large constructs and increase in DNA yields.

Genotype

F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 F80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697 galUgalK rpsL nupG (attL araC-PBAD-trfA250 bla attR) λ-

Features and Benefits

  • Cells that enable the most challenging library construction and screening.
  • Highest available transformation efficiencies for library construction and screening.
  • Optimized for high yields of DNA
  • Blue - white screening

Components

  • XLDNA V2 electrocompetent cells
  • pKanR transformation control DNA
  • recovery medium for cloning

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service