Merck
  • "PROTAC" modified dihydroquinolizinones (DHQs) that cause degradation of PAPD-5 and inhibition of hepatitis A virus and hepatitis B virus, in vitro.
"PROTAC" modified dihydroquinolizinones (DHQs) that cause degradation of PAPD-5 and inhibition of hepatitis A virus and hepatitis B virus, in vitro.
Bioorganic & medicinal chemistry letters (2024-03-02)
You Li, Nicky Hwang, Andrew Snedeker, Stanley M Lemon, Daisy Noe, Liren Sun, Jason A Clement, Tianlun Zhou, Liudi Tang, Timothy Block, Yanming Du
ABSTRACT
Dihydroquinolizinones (DHQs) that inhibit cellular polyadenylating polymerases 5 and 7 (PAPD5 & 7), such as RG7834, have been shown to inhibit both hepatitis A (HAV) and hepatitis B virus (HBV) in vitro and in vivo. In this report, we describe RG7834-based proteolysis-targeting chimeras (PROTACs), such as compound 12b, (6S)-9-((1-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-21-oxo-3,6,9,12,15,18-hexaoxa-22-azapentacosan-25-yl)oxy)-6-isopropyl-10-methoxy-2-oxo-6,7-dihydro-2H-pyrido[2,1-a]isoquinoline-3-carboxylic acid. The PROTAC DHQs described here inhibited an HAV reporter virus in vitro with an IC50 of 277 nM. Although the PROTAC DHQs were also inhibitory to HBV, their activities were substantially less potent against HBV in vitro, being in the 10 to 20 µM range, based on the reduction of HBsAg and HBV mRNA levels. Importantly, unlike RG7834, the incubation of cells in vitro with PROTAC DHQ 12b resulted in the degradation of PAPD5, as expected for a PROTAC compound, but curiously not PAPD7. PAPD5 polypeptide degradation was prevented when a proteasome inhibitor, epoxomicin, was used, indicating that proteasome mediated proteolysis was associated with the observed activities of 12b. Taken together, these data show that 12b is the first example of a PROTAC that suppresses both HAV and HBV that is based on a small molecule warhead. The possibility that it has mechanisms that differ from its parent compound, RG7834, and has clinical value, is discussed.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-PAPD7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution