Merck
  • Low-Dose Vertical Inhibition of the RAF-MEK-ERK Cascade Causes Apoptotic Death of KRAS Mutant Cancers.
Low-Dose Vertical Inhibition of the RAF-MEK-ERK Cascade Causes Apoptotic Death of KRAS Mutant Cancers.
Cell reports (2020-06-20)
Irem Ozkan-Dagliyan, J Nathaniel Diehl, Samuel D George, Antje Schaefer, Bjoern Papke, Kathleen Klotz-Noack, Andrew M Waters, Craig M Goodwin, Prson Gautam, Mariaelena Pierobon, Sen Peng, Thomas S K Gilbert, Kevin H Lin, Onur Dagliyan, Krister Wennerberg, Emanuel F Petricoin, Nhan L Tran, Shripad V Bhagwat, Ramon V Tiu, Sheng-Bin Peng, Laura E Herring, Lee M Graves, Christine Sers, Kris C Wood, Adrienne D Cox, Channing J Der
ABSTRACT
We address whether combinations with a pan-RAF inhibitor (RAFi) would be effective in KRAS mutant pancreatic ductal adenocarcinoma (PDAC). Chemical library and CRISPR genetic screens identify combinations causing apoptotic anti-tumor activity. The most potent combination, concurrent inhibition of RAF (RAFi) and ERK (ERKi), is highly synergistic at low doses in cell line, organoid, and rat models of PDAC, whereas each inhibitor alone is only cytostatic. Comprehensive mechanistic signaling studies using reverse phase protein array (RPPA) pathway mapping and RNA sequencing (RNA-seq) show that RAFi/ERKi induced insensitivity to loss of negative feedback and system failures including loss of ERK signaling, FOSL1, and MYC; shutdown of the MYC transcriptome; and induction of mesenchymal-to-epithelial transition. We conclude that low-dose vertical inhibition of the RAF-MEK-ERK cascade is an effective therapeutic strategy for KRAS mutant PDAC.
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
Cesium acetate, 99.9% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade