
Dopamine Related Compound B

Synonym(s):
3-Methoxytyramine hydrochloride, 3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing