
Caftaric acid

Synonym(s):
2-Caffeoyl-L-tartaric acid
Empirical Formula (Hill Notation):
C13H12O9
CAS Number:
Molecular Weight:
312.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4