![Boric acid gel 0.1-0.4 mm particle size](/deepweb/assets/sigmaaldrich/product/structures/310/992/6ce5c301-0542-4c07-b708-f42c4b527bb0/640/6ce5c301-0542-4c07-b708-f42c4b527bb0.png)
Boric acid gel
![Boric acid gel 0.1-0.4 mm particle size](/deepweb/assets/sigmaaldrich/product/structures/310/992/6ce5c301-0542-4c07-b708-f42c4b527bb0/640/6ce5c301-0542-4c07-b708-f42c4b527bb0.png)
Synonym(s):
3-Aminophenylboronic acid, polymer-bound
Empirical Formula (Hill Notation):
C10H12BNO3
CAS Number:
Molecular Weight:
205.02
Products
Brand
Formula Weight
Physical Form
Quality Segment
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing