Skip to Content
Merck
All Photos(1)

Documents

MABX8271

Sigma-Aldrich

Anti-Hepatitis B Surface Antigen (HBsAg) Antibody, (1mg)

clone 1, Chemicon®, from mouse

Synonym(s):

Hepatitis B surface antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1, monoclonal

species reactivity

hepatitis B virus (surface antigen), virus

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

The presence of HBsAg, a protein on the surface of HSV, indicates that a person is infectious. Hepatitis B virus surface antigen is tested for in the differential diagnosis of hepatitis and can be detected in high levels in serum during acute or chronic HBV infection. Early in the infection, this antigen may not be present and it may be undetectable later in the infection as it is being cleared by the host.

Specificity

Surface antigen of Hepatitis B virus (serotypes adw, ayw, adr)

Immunogen

Surface antigen of Hepatitis B virus (HBsAg)

Application

Detect HBsAg using mouse monoclonal Anti-HBsAg, Cat No MABX8271-1KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Infectious Diseases
Suggested antibody pairs MABX8271-1KC (labeling) and MABX8272-1KC (coating)

Quality

Detect HBsAg using mouse monoclonal Anti-HBsAg, Cat No MABX8271-1KC, validated for use in ELISA

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal antibody in buffer containing 10 mM Tris-HCl, pH 8.0, 0.05% NaN3

Storage and Stability

2-8C. For long-term storage, freeze at -20C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service