Skip to Content
Merck
All Photos(1)

Key Documents

B485-300DB

Rinse Fluid A

bottle of 300 mL, pkg of 4 units, square PETG bottle (500ml individually double wrapped with screw cap septum), sterile; aseptically filled

Synonym(s):

Fluid A for Sterility Testing, Rinse Fluid, Rinsing Solution Fluid A, Washing Fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

material

square PETG bottle (500ml individually double wrapped with screw cap septum)

Quality Level

Agency

EP 2.6.13
JP 4.06
USP 71

sterility

sterile; aseptically filled

form

liquid

packaging

bottle of 300 mL
pkg of 4 units

technique(s)

microbiological culture: suitable
sterility testing: suitable

pH range

6.9-7.3

application(s)

pharmaceutical

storage temp.

2-25°C

suitability

nonselective for bacteria
(

)

Looking for similar products? Visit Product Comparison Guide

General description

Fluid A is a washing solution for sterility testing of aqueous solutions or water-soluble pharmaceutical products using membrane filtration method.
Fluid A washing solution in general serves as a rinse buffer and can be used for an initial rinse of the filter membrane to inactivate any inhibitors, if present.

Application

Fluid A washing solution is a general rinse buffer and is compatible with most test samples. It is suitable for dissolving or diluting the test samples, reconstituting commercial microbes or as transport medium for a broad range of microorganisms.

Analysis Note

The appearance of the medium is clear and colorless to slightly yellowish.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service