- Previously undescribed vitamin D C-3 epimer occurs in substantial amounts in the blood of cats.
Previously undescribed vitamin D C-3 epimer occurs in substantial amounts in the blood of cats.
Journal of feline medicine and surgery (2017-11-28)
Megan C Sprinkle, Sarah E Hooper, Robert C Backus
PMID29172936
ABSTRACT
Objectives The aim of this report is to describe the identification of a novel vitamin D metabolite, a C-3, alpha-epimer of 25-hydroxycholecalciferol (3-epi-25(OH)D
MATERIALS