Merck
  • A phase 3, multicentre, randomized, double-blind, placebo-controlled, parallel-group study to evaluate the efficacy and safety of etelcalcetide (ONO-5163/AMG 416), a novel intravenous calcimimetic, for secondary hyperparathyroidism in Japanese haemodialysis patients.
A phase 3, multicentre, randomized, double-blind, placebo-controlled, parallel-group study to evaluate the efficacy and safety of etelcalcetide (ONO-5163/AMG 416), a novel intravenous calcimimetic, for secondary hyperparathyroidism in Japanese haemodialysis patients.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2017-01-07)
Masafumi Fukagawa, Keitaro Yokoyama, Takashi Shigematsu, Takashi Akiba, Akifumi Fujii, Takuto Kuramoto, Motoi Odani, Tadao Akizawa
ABSTRACT
Secondary hyperparathyroidism (SHPT) is a major complication associated with chronic kidney disease. We evaluated the efficacy and safety of etelcalcetide (ONO-5163/AMG 416), a novel intravenous calcimimetic, in Japanese haemodialysis patients with SHPT. In this phase 3, multicentre, randomized, double-blind, placebo-controlled, parallel-group study, etelcalcetide was administered three times per week at an initial dose of 5 mg, and subsequently adjusted to doses between 2.5 and 15 mg at 4-week intervals for 12 weeks. A total of 155 SHPT patients with serum intact parathyroid hormone (iPTH) levels ≥300 pg/mL were assigned to receive etelcalcetide (n = 78) or placebo (n = 77). The primary endpoint was the proportion of patients with decreased serum iPTH to the target range proposed by the Japanese Society for Dialysis Therapy (60-240 pg/mL). The major secondary endpoint was the proportion of patients with ≥30% reductions in serum iPTH from baseline. The proportion of patients meeting the primary endpoint was significantly higher for etelcalcetide (59.0%) versus placebo (1.3%). Similarly, the proportion of patients meeting the major secondary endpoint was significantly higher for etelcalcetide (76.9%) versus placebo (5.2%). Serum albumin-corrected calcium, phosphorus and intact fibroblast growth factor-23 levels were decreased in the etelcalcetide group. Nausea, vomiting and symptomatic hypocalcaemia were mild with etelcalcetide. Serious adverse events related to etelcalcetide were not observed. This study demonstrated the efficacy and safety of etelcalcetide. As the only available intravenous calcium-sensing receptor agonist, etelcalcetide is likely to provide a new treatment option for SHPT in haemodialysis patients.
Sigma-Aldrich
Human PLG (Plasminogen) ELISA Kit, for cell culture supernatants, plasma, and serum samples