Merck
Electrosynthesis of Bifunctional WS
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-04-28)
Shu Min Tan, Martin Pumera
ABSTRACT
A multitude of research into the application of transition-metal dichalcogenides as earth-abundant hydrogen evolution reaction (HER) electrocatalysts has been conducted. However, current synthetic methods generally deploy environmentally harmful chemicals and energy-consuming reaction conditions, despite the primary intent to attain renewable energy production. Here, the desirable properties of tungsten sulfide and reduced graphene oxide (rGO) have been combined and hybrid materials have been fabricated through simultaneous electrochemical reduction and synthesis, as a versatile and environmentally benign alternative to conventional fabrication techniques. Through concurrent studies of three rGO materials, the precursor of which was graphene oxide (GO), produced by Hummers, Staudenmaier, or Hofmann oxidation methods, the importance of the choice of oxidation method employed prior to the fabrication of the hybrid was shown. In this cardinal study, a mixed WS
Sigma-Aldrich
Ammonium tetrathiotungstate, ≥99.9% trace metals basis