Merck
  • Coordination polymer-derived nano-sized zinc ferrite with excellent performance in nitro-explosive detection.
Coordination polymer-derived nano-sized zinc ferrite with excellent performance in nitro-explosive detection.
Dalton transactions (Cambridge, England : 2003) (2017-08-16)
Debal Kanti Singha, Partha Mahata
ABSTRACT
Herein, a mixed metal coordination polymer, {(H
Sigma-Aldrich
2,5-Pyridinedicarboxylic acid, 98%
Sigma-Aldrich
TNP, ≥95% (HPLC)