Merck
  • RIPK3/Fas-Associated Death Domain Axis Regulates Pulmonary Immunopathology to Cryptococcal Infection Independent of Necroptosis.
RIPK3/Fas-Associated Death Domain Axis Regulates Pulmonary Immunopathology to Cryptococcal Infection Independent of Necroptosis.
Frontiers in immunology (2017-09-19)
Zhenzong Fa, Qun Xie, Wei Fang, Haibing Zhang, Haiwei Zhang, Jintao Xu, Weihua Pan, Jinhua Xu, Michal A Olszewski, Xiaoming Deng, Wanqing Liao
ABSTRACT
Fas-associated death domain (FADD) and receptor interacting protein kinase 3 (RIPK3) are multifunctional regulators of cell death and immune response. Using a mouse model of cryptococcal infection, the roles of FADD and RIPK3 in anti-cryptococcal defense were investigated. Deletion of RIPK3 alone led to increased inflammatory cytokine production in the
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine