Merck
  • Clinicopathological characteristics and genomic profile of primary sinonasal tract diffuse large B cell lymphoma (DLBCL) reveals gain at 1q31 and RGS1 encoding protein; high RGS1 immunohistochemical expression associates with poor overall survival in DLBCL not otherwise specified (NOS).
Clinicopathological characteristics and genomic profile of primary sinonasal tract diffuse large B cell lymphoma (DLBCL) reveals gain at 1q31 and RGS1 encoding protein; high RGS1 immunohistochemical expression associates with poor overall survival in DLBCL not otherwise specified (NOS).
Histopathology (2016-10-25)
Joaquim Carreras, Yara Y Kikuti, Sílvia Beà, Masashi Miyaoka, Shinichiro Hiraiwa, Haruka Ikoma, Ryoko Nagao, Sakura Tomita, David Martin-Garcia, Itziar Salaverria, Ai Sato, Akifumi Ichiki, Giovanna Roncador, Juan F Garcia, Kiyoshi Ando, Elias Campo, Naoya Nakamura
ABSTRACT
We aimed to define the clinicopathological characteristics of 29 primary sinonasal diffuse large B cell lymphoma (DLBCL In the training set, 82% had a non-germinal center B-cell-like (Hans' Classifier) (non-GCB) phenotype and 18% were Epstein-Barr virus-encoded small RNAs (EBER) DLBCL