Merck
  • Real Time-NIR/MIR-Photorheology: A Versatile Tool for the in Situ Characterization of Photopolymerization Reactions.
Real Time-NIR/MIR-Photorheology: A Versatile Tool for the in Situ Characterization of Photopolymerization Reactions.
Analytical chemistry (2017-04-07)
Christian Gorsche, Reghunathan Harikrishna, Stefan Baudis, Patrick Knaack, Branislav Husar, Joerg Laeuger, Helmuth Hoffmann, Robert Liska
ABSTRACT
In photopolymerization reactions, mostly multifunctional monomers are employed, as they ensure fast reaction times and good final mechanical properties of the cured materials. Drawing conclusions about the influence of the components and curing conditions on the mechanical properties of the subsequently formed insoluble networks is challenging. Therefore, an in situ observation of chemical and mechanical characteristics during the photopolymerization reaction is desired. By coupling of an infrared spectrometer with a photorheometer, a broad spectrum of different photopolymerizable formulations can be analyzed during the curing reaction. The rheological information (i.e., time to gelation, final modulus, shrinkage force) can be derived from a parallel plate rheometer equipped with a UV- and IR-translucent window (glass for NIR and CaF
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
1,6-Hexanediol diacrylate, technical grade, 80%