- Activity of anti-inflammatory, analgesic and antigenotoxic of the aqueous flower extracts of Opuntia microdasys Lem.Pfeiff.
Activity of anti-inflammatory, analgesic and antigenotoxic of the aqueous flower extracts of Opuntia microdasys Lem.Pfeiff.
The aim of this study was to evaluate the analgesic and the anti-inflammatory activity of Opuntia microdasys at post flowering stage, F3 (OMF3) in rat and, in other hand, its antigenotoxic effects by the Allium cepa test. OMF3 extracts were screened for activity of analgesic and anti-inflammatory using, respectively, the acetic acid writhing test in mice and the carrageenan-induced paw oedema assay in rats. The antigenotoxic has been evaluated by A. cepa test. OMF3 extracts showed a higher analgesic and anti-inflammatory activity at 100 mg/kg (72.03% and 70.11%) as determined by the tests of acetic acid-induced writhing and carrageenan-induced oedema, respectively. Furthermore, the OMF3 aqueous extracts have a preventive antimutagenic potential, at lower concentration (EC Based on this study, the flower of O. microdasys at post flowering stage may be use as an analgesic, anti-inflammatory and antimutagenic agents.