Merck
  • From common to rare Zingiberaceae plants - A metabolomics study using GC-MS.
From common to rare Zingiberaceae plants - A metabolomics study using GC-MS.
Phytochemistry (2017-05-13)
Gina B Barbosa, Nirupama S Jayasinghe, Siria H A Natera, Ellen D Inutan, Nonita P Peteros, Ute Roessner
ABSTRACT
Zingiberaceae plants, commonly known as gingers, have been popular for their medicinal and culinary uses since time immemorial. In spite of their numerous health-promoting applications, many Zingiberaceae plants still receive no scientific attention. Moreover, existing reports mostly focused only on the Zingiberaceae rhizomes. Here, untargeted metabolite profiling using Gas Chromatography - Mass Spectrometry (GC-MS) was used to compare the metabolic composition of leaves and rhizomes of the more common gingers, Zingiber officinale Rosc. (ZO), Curcuma longa L. (CL), and Etlingera elatior (Jack) R.M. Smith (EE), and the rare gingers, Amomum muricarpum Elmer (AM), Etlingera philippinensis (Ridl.) R.M. Smith (EP), and Hornstedtia conoidea Ridl. (HC). Principal component analysis (PCA) demonstrated that different species show substantial chemical differentiation and revealed potential markers among the different Zingiberaceae plants. Interestingly, the leaves of AM, CL, EE, EP, and HC had significantly higher levels of chlorogenic acid than ZO. Moreover, rhizomes of EP and HC were found to contain significantly higher levels of amino acids than ZO. Sugars and organic acids were generally less abundant in ZO leaves and rhizomes than in the other gingers. The leaves of EP and rhizomes of AM were found most similar to the leaves and rhizomes of common gingers, respectively. Results of this study provide significant baseline information on assessing the possible usage of the leaves of common gingers and further propagation and exploration of EP and AM. This study, being the first metabolomics report on rare plants such as AM, EP and HC, affirms the usefulness of untargeted metabolite profiling in exploring under-investigated plants.
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoic acid, 99%