Merck
  • Characterization of Mesocarp and Kernel Lipids from Elaeis guineensis Jacq., Elaeis oleifera [Kunth] Cortés, and Their Interspecific Hybrids.
Characterization of Mesocarp and Kernel Lipids from Elaeis guineensis Jacq., Elaeis oleifera [Kunth] Cortés, and Their Interspecific Hybrids.
Journal of agricultural and food chemistry (2017-04-25)
Veronika M Lieb, Margarete R Kerfers, Amrei Kronmüller, Patricia Esquivel, Amancio Alvarado, Víctor M Jiménez, Hans-Georg Schmarr, Reinhold Carle, Ralf M Schweiggert, Christof B Steingass
ABSTRACT
Morphological traits, total lipid contents, and fatty acid profiles were assessed in fruits of several accessions of Elaeis oleifera [Kunth] Cortés, Elaeis guineensis Jacq., and their interspecific hybrids. The latter featured the highest mesocarp-to-fruit ratios (77.9-78.2%). The total lipid contents of both E. guineensis mesocarp and kernel were significantly higher than for E. oleifera accessions. Main fatty acids comprised C16:0, C18:1n9, and C18:2n6 in mesocarp and C12:0, C14:0, and C18:1n9 in kernels. E. oleifera samples were characterized by higher proportions of unsaturated long-chain fatty acids. Saturated medium-chain fatty acids supported the clustering of E. guineensis kernels in multivariate statistics. Hybrid mesocarp lipids had an intermediate fatty acid composition, whereas their kernel lipids resembled those of E. oleifera genotypes. Principal component analysis based on lipid contents and proportions of individual fatty acids permitted clear-cut distinction of E. oleifera, E. guineensis, and their hybrids.
Sigma-Aldrich
Methyl trans-vaccenate, ≥99% (capillary GC), liquid