Merck
  • Chiral separation of new sulfonamide derivatives and evaluation of their enantioselective affinity for human carbonic anhydrase II by microscale thermophoresis and surface plasmon resonance.
Chiral separation of new sulfonamide derivatives and evaluation of their enantioselective affinity for human carbonic anhydrase II by microscale thermophoresis and surface plasmon resonance.
Journal of pharmaceutical and biomedical analysis (2017-01-23)
Tiphaine Rogez-Florent, Catherine Foulon, Anne-Sophie Drucbert, Nadège Schifano, Perrine Six, Stéphanie Devassine, Patrick Depreux, Pierre-Marie Danzé, Laurence Goossens, Cécile Danel, Jean-François Goossens
ABSTRACT
The aim of this study was to develop a method combining chiral separation and biophysical techniques to evaluate the enantioselective affinity of original sulfonamide derivatives towards their therapeutic target, the human carbonic anhydrase II (hACII). The first step consisted in the preparation of the enantiomers by chromatographic separation. The performances of HPLC and Supercritical Fluid Chromatography (SFC) were studied at the analytical scale by optimization of various experimental conditions using adsorbed polysaccharide chiral stationary phases (amylose AD-H and cellulose OD-H). Since SFC allowed obtaining higher enantioresolutions per time unit, it was selected for the semi-preparative scale and successfully used to isolate each enantiomer with a satisfactory enantiomeric purity (>98%). Secondly, microscale thermophoresis (MST) method and surface plasmon resonance (SPR) used as reference method were developed to measure potential enantioselective affinities of these enantiomers towards the hACII. The optimizations of both methods were performed using a reference compound, i.e. acetazolamide, which affinity for hCAII has previously been demonstrated. For all compounds, K
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)