Merck
Reproducibility of urinary biomarkers in multiple 24-h urine samples.
The American journal of clinical nutrition (2017-01-05)
Qi Sun, Kimberly A Bertrand, Adrian A Franke, Bernard Rosner, Gary C Curhan, Walter C Willett
ABSTRACT
Limited knowledge regarding the reproducibility of biomarkers in 24-h urine samples has hindered the collection and use of the samples in epidemiologic studies. We aimed to evaluate the reproducibility of various markers in repeat 24-h urine samples. We calculated intraclass correlation coefficients (ICCs) of biomarkers measured in 24-h urine samples that were collected in 3168 participants in the NHS (Nurses' Health Study), NHSII (Nurses' Health Study II), and Health Professionals Follow-Up Study. In 742 women with 4 samples each collected over the course of 1 y, ICCs for sodium were 0.32 in the NHS and 0.34 in the NHSII. In 2439 men and women with 2 samples each collected over 1 wk to ≥1 mo, the ICCs ranged from 0.33 to 0.68 for sodium at various intervals between collections. The urinary excretion of potassium, calcium, magnesium, phosphate, sulfate, and other urinary markers showed generally higher reproducibility (ICCs >0.4). In 47 women with two 24-h urine samples, ICCs ranged from 0.15 (catechin) to 0.75 (enterolactone) for polyphenol metabolites. For phthalates, ICCs were generally ≤0.26 except for monobenzyl phthalate (ICC: 0.55), whereas the ICC was 0.39 for bisphenol A (BPA). We further estimated that, for the large majority of the biomarkers, the mean of three 24-h urine samples could provide a correlation of ≥0.8 with true long-term urinary excretion. These data suggest that the urinary excretion of various biomarkers, such as minerals, electrolytes, most polyphenols, and BPA, is reasonably reproducible in 24-h urine samples that are collected within a few days or ≤1 y. Our findings show that three 24-h samples are sufficient for the measurement of long-term exposure status in epidemiologic studies.
Sigma-Aldrich
Enterodiol, ≥95.0% (HPLC)
Sigma-Aldrich
Enterolactone, ~95% (HPLC)