Merck
  • IRF5 regulates lung macrophages M2 polarization during severe acute pancreatitis
IRF5 regulates lung macrophages M2 polarization during severe acute pancreatitis
World journal of gastroenterology (2016-11-30)
Kang Sun, Song-Bing He, Jian-Guo Qu, Sheng-Chun Dang, Ji-Xiang Chen, Ai-Hua Gong, Rong Xie, Jian-Xin Zhang
ABSTRACT
To investigate the role of interferon regulatory factor 5 (IRF5) in reversing polarization of lung macrophages during severe acute pancreatitis (SAP) A mouse SAP model was established by intraperitoneal (ip) injections of 20 μg/kg body weight caerulein. Pathological changes in the lung were observed by hematoxylin and eosin staining. Lung macrophages were isolated from bronchoalveolar lavage fluid. The quantity and purity of lung macrophages were detected by fluorescence-activated cell sorting and evaluated by real-time polymerase chain reaction (RT-PCR). They were treated with IL-4/IRF5 specific siRNA (IRF5 siRNA) to reverse their polarization and were evaluated by detecting markers expression of M1/M2 using RT-PCR. SAP associated acute lung injury (ALI) was induced successfully by ip injections of caerulein, which was confirmed by histopathology. Lung macrophages expressed high levels of IRF5 as M1 phenotype during the early acute pancreatitis stages. Reduction of IRF5 expression by IRF5 siRNA reversed the action of macrophages from M1 to M2 phenotype Treatment with IRF5 siRNA can reverse the pancreatitis-induced activation of lung macrophages from M1 phenotype to M2 phenotype in SAP associated with ALI.
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF5