Merck
  • Targeting Homologous Recombination by Pharmacological Inhibitors Enhances the Killing Response of Glioblastoma Cells Treated with Alkylating Drugs.
Targeting Homologous Recombination by Pharmacological Inhibitors Enhances the Killing Response of Glioblastoma Cells Treated with Alkylating Drugs.
Molecular cancer therapeutics (2016-11-04)
Nancy Berte, Andrea Piée-Staffa, Nadine Piecha, Mengwan Wang, Kerstin Borgmann, Bernd Kaina, Teodora Nikolova
ABSTRACT
Malignant gliomas exhibit a high level of intrinsic and acquired drug resistance and have a dismal prognosis. First- and second-line therapeutics for glioblastomas are alkylating agents, including the chloroethylating nitrosoureas (CNU) lomustine, nimustine, fotemustine, and carmustine. These agents target the tumor DNA, forming O